Order Entry
Czech Republic
ContactUsLinkComponent
3-(Dimethylamino)-1-[3-(trifluoromethoxy)phenyl]prop-2-en-1-one
Irritant
3-(Dimethylamino)-1-[3-(trifluoromethoxy)phenyl]prop-2-en-1-one
Catalog # APOSPC300780-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(Dimethylamino)-1-[3-(trifluoromethoxy)phenyl]prop-2-en-1-one
Catalog # APOSPC300780-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.224 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:154258-46-5
Formula:C₁₂H₁₂F₃NO₂
MDL Number:MFCD26939178