2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:852952-20-6
:
Formula:C₁₃H₇ClF₃NO₃
MDL Number:MFCD03644663
:317.648 g/mol
Storage Temperature:Ambient
Frequently Bought Together