Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Iodo-2-methyl-6-(trifluoromethyl)pyridine
3-Iodo-2-methyl-6-(trifluoromethyl)pyridine
3-Iodo-2-methyl-6-(trifluoromethyl)pyridine
3-Iodo-2-methyl-6-(trifluoromethyl)pyridine
APOSPC300768-1G
:
Apollo Scientific
:
3-Iodo-2-methyl-6-(trifluoromethyl)pyridine
APOSPC300768-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
287.021 g/mol
:
GHS07
:
Ambient
:
944317-26-4
:
C₇H₅F₃IN
:
MFCD27934112