Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-[(Dimethylamino)methylene]-6-fluoroindan-1-one
2-[(Dimethylamino)methylene]-6-fluoroindan-1-one
2-[(Dimethylamino)methylene]-6-fluoroindan-1-one
2-[(Dimethylamino)methylene]-6-fluoroindan-1-one
APOSPC300746-100MG
:
Apollo Scientific
:
2-[(Dimethylamino)methylene]-6-fluoroindan-1-one
APOSPC300746-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
205.228 g/mol
:
GHS07
:
Ambient
:
1427462-76-7
:
C₁₂H₁₂FNO
:
MFCD19442756