
3-Chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Catalog # APOSPC300739-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:264.588 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1237838-81-1
Formula:C₉H₄ClF₃N₂O₂
MDL Number:MFCD17168283