
3-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Catalog # APOSPC300738-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:309.039 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1355171-41-3
Formula:C₉H₄BrF₃N₂O₂
MDL Number:MFCD17168281