{5-[3,5-Di(trifluoromethyl)phenyl]-2-furyl}methanol
Catalog # APOSPC300695-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:310.192 g/mol
Storage Temperature:Ambient
CAS Number:424803-19-0
Formula:C₁₃H₈F₆O₂
MDL Number:MFCD00052573