4-{2-[5-(Trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-4-yl}benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₄H₈F₃N₃O₂S
MDL Number:MFCD11846269
:339.292 g/mol
Storage Temperature:Ambient
Frequently Bought Together