4-Methyl-2-[5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-5-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₉H₆F₃N₃O₂S
MDL Number:MFCD11846268
:277.223 g/mol
Storage Temperature:Ambient
Frequently Bought Together