3-{4-[4-(Trifluoromethyl)pyrimidin-2-yl]homopiperazin-1-yl}propanoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:874802-36-5
:
Formula:C₁₃H₁₇F₃N₄O₂
MDL Number:MFCD05664937
:318.295 g/mol
Storage Temperature:Ambient
Frequently Bought Together