Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-[4-(Bromomethyl)phenoxy]-3-fluorobenzene
1-[4-(Bromomethyl)phenoxy]-3-fluorobenzene
1-[4-(Bromomethyl)phenoxy]-3-fluorobenzene
1-[4-(Bromomethyl)phenoxy]-3-fluorobenzene
APOSPC300615-1G
:
Apollo Scientific
:
1-[4-(Bromomethyl)phenoxy]-3-fluorobenzene
APOSPC300615-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
281.12 g/mol
:
Ambient
:
1355171-43-5
:
C₁₃H₁₀BrFO
:
MFCD16619596