Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
[2-(2-Fluorophenoxy)phenyl]methanol
[2-(2-Fluorophenoxy)phenyl]methanol
[2-(2-Fluorophenoxy)phenyl]methanol
APOSPC300606-1G
:
Apollo Scientific
:
[2-(2-Fluorophenoxy)phenyl]methanol
APOSPC300606-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
218.224 g/mol
:
Ambient
:
81890-62-2
:
C₁₃H₁₁FO₂
:
MFCD04108476