4-[4-(Trifluoromethoxy)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:155431-35-9
:
Formula:C₉H₆F₃N₃O₂
MDL Number:MFCD04125794
:245.158 g/mol
Storage Temperature:Ambient
Frequently Bought Together