
Ethyl 3,8-dichloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSPC300582-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:327.087 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1355171-49-1
Formula:C₁₁H₇Cl₂F₃N₂O₂
MDL Number:MFCD17168289