
Ethyl 3-chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSPC300577-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:292.642 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1355171-67-3
Formula:C₁₁H₈ClF₃N₂O₂
MDL Number:MFCD17168282