Order Entry
Czech Republic
ContactUsLinkComponent
N-Methyl-3-(trifluoromethyl)pyridin-2-amine > 95%
Irritant
N-Methyl-3-(trifluoromethyl)pyridin-2-amine > 95%
Catalog # APOSPC300563-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-Methyl-3-(trifluoromethyl)pyridin-2-amine > 95%
Catalog # APOSPC300563-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.139 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1036584-14-1
Formula:C₇H₇F₃N₂
MDL Number:MFCD11118894