Order Entry
Czech Republic
ContactUsLinkComponent
2-(But-1-yl)-5-(trifluoromethyl)-1,3-benzoxazole
2-(But-1-yl)-5-(trifluoromethyl)-1,3-benzoxazole
Catalog # APOSPC300513-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(But-1-yl)-5-(trifluoromethyl)-1,3-benzoxazole
Catalog # APOSPC300513-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.225 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₂F₃NO
MDL Number:MFCD22628770