Order Entry
Czech Republic
ContactUsLinkComponent
2-Ethyl-5-(trifluoromethyl)-1,3-benzoxazole
2-Ethyl-5-(trifluoromethyl)-1,3-benzoxazole
Catalog # APOSPC300511-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Ethyl-5-(trifluoromethyl)-1,3-benzoxazole
Catalog # APOSPC300511-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.172 g/mol
Storage Temperature:Ambient
CAS Number:1267428-36-3
Formula:C₁₀H₈F₃NO
MDL Number:MFCD19375803