
1,1,1-Trifluoropropan-2-amine hydrochloride
Catalog # APOSPC2929-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:149.543 g/mol
Melting Pt:218…221 °C (subl.)
:GHS07
Storage Temperature:Ambient
CAS Number:2968-32-3
:421-49-8
Formula:C₃H₆F₃N·ClH
MDL Number:MFCD04972986