Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-7-(trifluoromethoxy)quinoline
4-Chloro-7-(trifluoromethoxy)quinoline
Catalog # APOSPC28306-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-7-(trifluoromethoxy)quinoline
Catalog # APOSPC28306-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.601 g/mol
Storage Temperature:Refrigerator
CAS Number:40516-31-2
Formula:C₁₀H₅ClF₃NO
MDL Number:MFCD08063185