
3-Benzyl-8-(trifluoromethyl)-3,4-dihydropyrimido[2,1-b][1,3,5]thiadiazin-6(2H)-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1426894-90-7
:
Formula:C₁₄H₁₂F₃N₃OS
MDL Number:MFCD31812103
:327.325 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together