
Ethyl 2-(6-oxo-8-(trifluoromethyl)pyrimido[2,1-b][1,3,5]thiadiazin-3(2H,4H,6H)-yl)acetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1622837-13-1
:
Formula:C₁₁H₁₂F₃N₃O₃S
MDL Number:MFCD31812102
:323.291 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together