
N-((Dimethylamino)methylene)-2,4-difluorobenzamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2379917-16-3
:
Formula:C₁₀H₁₀F₂N₂O
MDL Number:MFCD31812108
:212.196 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together