
3-Chloro-N'-hydroxy-5-(trifluoromethyl)picolinimidamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:303997-55-9
:
Formula:C₇H₅ClF₃N₃O
MDL Number:MFCD00231716
:239.582 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together