
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1823188-42-6
:
Formula:C₉H₄ClF₃N₂O₂
MDL Number:MFCD12406878
:264.588 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together