Order Entry
Czech Republic
ContactUsLinkComponent
4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzaldehyde 97%
4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzaldehyde 97%
Catalog # APOSPC28233-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzaldehyde 97%
Catalog # APOSPC28233-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.181 g/mol
Storage Temperature:Ambient
CAS Number:1393125-27-3
Formula:C₁₁H₇F₃N₂O
MDL Number:MFCD19586808