Order Entry
Czech Republic
ContactUsLinkComponent
8-Chloro-3-(difluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech
8-Chloro-3-(difluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech
Catalog # APOSPC28192-1G
Supplier:  Apollo Scientific
CAS Number:  
8-Chloro-3-(difluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech
Catalog # APOSPC28192-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :271.575 g/mol
Storage Temperature:Ambient
CAS Number:478248-92-9
Formula:C₈H₃ClF₅N₃
MDL Number:MFCD02082839