Order Entry
Czech Republic
ContactUsLinkComponent
2-(2-Fluorophenyl)-2-(morpholin-4-yl)acetonitrile, tech
2-(2-Fluorophenyl)-2-(morpholin-4-yl)acetonitrile, tech
Catalog # APOSPC28170-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Fluorophenyl)-2-(morpholin-4-yl)acetonitrile, tech
Catalog # APOSPC28170-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.243 g/mol
Storage Temperature:Ambient
CAS Number:66549-00-6
Formula:C₁₂H₁₃FN₂O
MDL Number:MFCD00173114