Methyl 3-(4-fluorophenyl)-4-oxo-3,4-dihydrophthalazine-1-carboxylate, tech
Catalog # APOSPC28163-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:298.269 g/mol
Storage Temperature:Ambient
CAS Number:339021-34-0
Formula:C₁₆H₁₁FN₂O₃
MDL Number:MFCD00138620