1-[2-Amino-4,6-bis(trifluoromethyl)phenyl]ethan-1-one, tech
Catalog # APOSPC28128-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.159 g/mol
Storage Temperature:Ambient
CAS Number:882747-78-6
Formula:C₁₀H₇F₆NO
MDL Number:MFCD08281871