Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 5,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, tech
Ethyl 5,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, tech
Catalog # APOSPC28114-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, tech
Catalog # APOSPC28114-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :253.202 g/mol
Storage Temperature:Ambient
CAS Number:185011-67-0
Formula:C₁₂H₉F₂NO₃
MDL Number:MFCD00109441