Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylate, tech
Catalog # APOSPC28108-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:270.258 g/mol
Storage Temperature:Ambient
CAS Number:866041-17-0
Formula:C₁₅H₁₁FN₂O₂
MDL Number:MFCD05256131