3-Chloro-2-(pyridine-3-carbonyl)-5-(trifluoromethyl)pyridine, tech
Catalog # APOSPC28089-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.637 g/mol
Storage Temperature:Ambient
CAS Number:338953-55-2
Formula:C₁₂H₆ClF₃N₂O
MDL Number:MFCD00244682