(2E)-3-(Dimethylamino)-1-(2-fluorophenyl)prop-2-en-1-one, tech
Catalog # APOSPC28088-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:193.217 g/mol
Storage Temperature:Ambient
CAS Number:138716-19-5
Formula:C₁₁H₁₂FNO
MDL Number:MFCD00172154