4-[3-(Trifluoromethyl)phenyl]pyrimidin-2-amine, tech
Catalog # APOSPC28080-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:239.197 g/mol
Storage Temperature:Ambient
CAS Number:866019-76-3
Formula:C₁₁H₈F₃N₃
MDL Number:MFCD03407835