5-[3-(Trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
Catalog # APOSPC28079-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:288.227 g/mol
Storage Temperature:Ambient
CAS Number:866019-49-0
Formula:C₁₄H₇F₃N₄
MDL Number:MFCD04125471