1-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propan-2-one, tech
Catalog # APOSPC28033-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.197 g/mol
Storage Temperature:Ambient
CAS Number:321980-88-5
Formula:C₁₁H₉F₃N₂O
MDL Number:MFCD01419489