Order Entry
Czech Republic
ContactUsLinkComponent
3-[(3-Fluorophenyl)methyl]-2,4-dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile, tech
3-[(3-Fluorophenyl)methyl]-2,4-dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile, tech
Catalog # APOSPC28031-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[(3-Fluorophenyl)methyl]-2,4-dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile, tech
Catalog # APOSPC28031-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :280.3 g/mol
Storage Temperature:Ambient
CAS Number:477890-40-7
Formula:C₁₆H₁₃FN₄
MDL Number:MFCD02571880