Order Entry
Czech Republic
ContactUsLinkComponent
 
4-[(2-Chloro-4-fluorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech
Catalog # APOSPC28029-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[(2-Chloro-4-fluorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech
Catalog # APOSPC28029-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :293.742 g/mol
Storage Temperature:Ambient
CAS Number:477889-61-5
Formula:C₁₁H₁₃ClFNO₃S
MDL Number:MFCD02102594