Order Entry
Czech Republic
ContactUsLinkComponent
3-(1,1-Difluorobut-3-en-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech
3-(1,1-Difluorobut-3-en-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech
Catalog # APOSPC28028-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(1,1-Difluorobut-3-en-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech
Catalog # APOSPC28028-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :277.193 g/mol
Storage Temperature:Ambient
CAS Number:477889-55-7
Formula:C₁₁H₈F₅N₃
MDL Number:MFCD02571332