2-[3-(Trifluoromethyl)phenoxy]ethan-1-amine hydrochloride 95%
Catalog # APOSPC28023-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:241.638 g/mol
Storage Temperature:Ambient
CAS Number:104774-96-1
Formula:C₉H₁₀F₃NO·ClH
MDL Number:MFCD00793842