Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate 95%
Ethyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate 95%
Catalog # APOSPC28017-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate 95%
Catalog # APOSPC28017-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.223 g/mol
Storage Temperature:Ambient
CAS Number:82791-95-5
Formula:C₁₂H₁₁F₃N₂O₂
MDL Number:MFCD01956277