3-(2-Fluorophenoxy)butan-2-one, tech
Catalog # APOSPC27987-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.192 g/mol
Storage Temperature:Ambient
CAS Number:30343-27-2
Formula:C₁₀H₁₁FO₂
MDL Number:MFCD00243594