Order Entry
Czech Republic
ContactUsLinkComponent
(3Z)-3-(1-{[2-(Trifluoromethyl)phenyl]amino}ethylidene)oxolan-2-one, tech
(3Z)-3-(1-{[2-(Trifluoromethyl)phenyl]amino}ethylidene)oxolan-2-one, tech
Catalog # APOSPC27981-1G
Supplier:  Apollo Scientific
CAS Number:  
(3Z)-3-(1-{[2-(Trifluoromethyl)phenyl]amino}ethylidene)oxolan-2-one, tech
Catalog # APOSPC27981-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :271.235 g/mol
Storage Temperature:Ambient
CAS Number:685108-57-0
Formula:C₁₃H₁₂F₃NO₂
MDL Number:MFCD03617585