Order Entry
Czech Republic
ContactUsLinkComponent
(2E)-3-(Dimethylamino)-1-[3-fluoro-5-(trifluoromethyl)phenyl]prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-[3-fluoro-5-(trifluoromethyl)phenyl]prop-2-en-1-one, tech
Catalog # APOSPC27964-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-3-(Dimethylamino)-1-[3-fluoro-5-(trifluoromethyl)phenyl]prop-2-en-1-one, tech
Catalog # APOSPC27964-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :261.215 g/mol
Storage Temperature:Ambient
CAS Number:154258-42-1
Formula:C₁₂H₁₁F₄NO
MDL Number:MFCD06496095