Order Entry
Czech Republic
ContactUsLinkComponent
N-[1-(Hydrazinocarbonyl)-3-(methylthio)prop-1-yl]-4-(trifluoromethyl)benzamide 97%
N-[1-(Hydrazinocarbonyl)-3-(methylthio)prop-1-yl]-4-(trifluoromethyl)benzamide 97%
Catalog # APOSPC2781-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[1-(Hydrazinocarbonyl)-3-(methylthio)prop-1-yl]-4-(trifluoromethyl)benzamide 97%
Catalog # APOSPC2781-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :335.345 g/mol
Melting Pt:181…184 °C
Storage Temperature:Ambient
CAS Number:209056-84-8
Formula:C₁₃H₁₆F₃N₃O₂S
MDL Number:MFCD00174305