
2,6-Difluorophenylhydrazine hydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:502496-26-6
:
Formula:C₆H₆F₂N₂·ClH
MDL Number:MFCD03094169
:180.583 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together