Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Phenylthio)-5-(trifluoromethyl)benzoic acid
2-(Phenylthio)-5-(trifluoromethyl)benzoic acid
2-(Phenylthio)-5-(trifluoromethyl)benzoic acid
2-(Phenylthio)-5-(trifluoromethyl)benzoic acid
APOSPC2524-1G
:
Apollo Scientific
:
2-(Phenylthio)-5-(trifluoromethyl)benzoic acid
APOSPC2524-1G
:
Apollo Scientific
:
Typ balení:
Bottle
Bal.:
1 g
Sekvence:
3047
:
:
298.28 g/mol
:
153…154 °C
:
GHS07
:
Ambient
:
52548-96-6
:
C₁₄H₉F₃O₂S
:
MFCD00069137