

1,1-Difluoropropane (FC-272fb) 97%
Catalog # APOSPC2405-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Cylinder
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:80.077 g/mol
Boiling Pt:44415 °C
Storage Temperature:Ambient
UN:3161
ADR:2.1,
:GHS07
:GHS04
CAS Number:430-61-5
Formula:C₃H₆F₂
MDL Number:MFCD09258968