
4-(Difluoromethoxy)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:351003-34-4
:
Formula:C₇H₅ClF₂O₃S
MDL Number:MFCD03094156
:242.628 g/mol
Boiling Pt:265…266 °C
Density:1.523 g/cm³ (25 °C)
Flash Pt:93 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together